Assay Details
| Gene Symbol: pkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016470.3 | NP_001016470.1 | 8 - 9 | 1253 | 57 | |
| NM_001195712.1 | NP_001182641.1 | 8 - 9 | 1178 | 57 | ||
| GenBank mRNA | BC080927.1 | - | 7 - 8 | 1149 | 57 | |
| BC161037.1 | - | 8 - 9 | 1219 | 57 | ||
| BC170795.1 | - | 8 - 9 | 1180 | 57 | ||
| BC170799.1 | - | 8 - 9 | 1180 | 57 | ||
| CR760921.2 | - | 8 - 9 | 1186 | 57 | ||
More Information
Set Membership: |
|

pkm
Genomic Map