Assay Details
| Gene Symbol: cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016479.2 | NP_001016479.1 | 1 - 2 | 86 | 71 | |
| XM_012961095.2 | XP_012816549.1 | 2 - 3 | 175 | 71 | ||
| XM_012961096.1 | XP_012816550.1 | 2 - 3 | 276 | 71 | ||
| GenBank mRNA | BC135405.1 | - | 2 - 3 | 144 | 71 | |
| CR760893.2 | - | 1 - 2 | 86 | 71 | ||
More Information
Set Membership: |
|

cox4i1
Genomic Map