Assay Details
| Gene Symbol: mrps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016526.2 | NP_001016526.1 | 1 - 2 | 107 | 69 | |
| XM_012959027.1 | XP_012814481.1 | 2 - 3 | 182 | 69 | ||
| GenBank mRNA | CR926198.2 | - | 1 - 2 | 107 | 69 | |
More Information
Set Membership: |
|

mrps24
Genomic Map