Assay Details
| Gene Symbol: aldh3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016537.2 | NP_001016537.1 | 3 - 4 | 521 | 68 | |
| XM_012956976.2 | XP_012812430.1 | 4 - 5 | 651 | 68 | ||
| XM_012956977.2 | XP_012812431.1 | 4 - 5 | 619 | 68 | ||
| XM_012956978.2 | XP_012812432.1 | 4 - 5 | 650 | 68 | ||
| XM_012956979.2 | XP_012812433.1 | 4 - 5 | 630 | 68 | ||
| XM_012956980.2 | XP_012812434.1 | 4 - 5 | 601 | 68 | ||
| GenBank mRNA | BC091032.1 | - | 3 - 4 | 521 | 68 | |
| CR760779.2 | - | 3 - 4 | 521 | 68 | ||
More Information
Set Membership: |
|

aldh3a2
Genomic Map