Assay Details
| Gene Symbol: mfsd14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016552.2 | NP_001016552.1 | 11 - 12 | 1148 | 60 | |
| XM_012961107.2 | XP_012816561.1 | 11 - 12 | 1262 | 60 | ||
| GenBank mRNA | BC170818.1 | - | 10 - 11 | 1141 | 60 | |
| BC170820.1 | - | 10 - 11 | 1141 | 60 | ||
| CR760718.2 | - | 11 - 12 | 1148 | 60 | ||
More Information
Set Membership: |
|

mfsd14a
Genomic Map