Assay Details
| Gene Symbol: pold3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016556.2 | NP_001016556.1 | 445 | 85 | ||
| GenBank mRNA | CR760707.2 | - | 445 | 85 | ||
| CU075577.1 | - | 457 | 85 | |||
More Information
Set Membership: |
|

pold3
Genomic Map