Assay Details
| Gene Symbol: psmd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016559.2 | NP_001016559.1 | 3 - 4 | 454 | 82 | |
| GenBank mRNA | BC160446.1 | - | 3 - 4 | 442 | 82 | |
| BC167121.1 | - | 2 - 3 | 1276 | 82 | ||
| BC170825.1 | - | 3 - 4 | 454 | 82 | ||
| BC170829.1 | - | 3 - 4 | 454 | 82 | ||
| CR760701.2 | - | 3 - 4 | 454 | 82 | ||
More Information
Set Membership: |
|

psmd9
Genomic Map