Assay Details
| Gene Symbol: ap1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016579.3 | NP_001016579.1 | 2 - 3 | 249 | 82 | |
| GenBank mRNA | AL866501.2 | - | 2 - 3 | 250 | 82 | |
| AL883278.2 | - | 2 - 3 | 246 | 82 | ||
| CR760637.2 | - | 1 - 2 | 189 | 82 | ||
| CR926233.1 | - | 1 - 2 | 92 | 82 | ||
More Information
Set Membership: |
|

ap1s3
Genomic Map