Assay Details
| Gene Symbol: rpp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016584.2 | NP_001016584.1 | 10 - 11 | 686 | 65 | |
| XM_012966612.2 | XP_012822066.1 | 10 - 11 | 854 | 65 | ||
| GenBank mRNA | CR760621.2 | - | 10 - 11 | 686 | 65 | |
More Information
Set Membership: |
|

rpp30
Genomic Map