Assay Details
| Gene Symbol: immp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016589.2 | NP_001016589.1 | 3 - 4 | 246 | 68 | |
| GenBank mRNA | BC170762.1 | - | 2 - 3 | 230 | 68 | |
| BC170766.1 | - | 2 - 3 | 230 | 68 | ||
| CR760609.2 | - | 3 - 4 | 246 | 68 | ||
More Information
Set Membership: |
|

immp1l
Genomic Map