Assay Details
| Gene Symbol: atg4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016619.2 | NP_001016619.1 | 4 - 5 | 333 | 64 | |
| XM_018096264.1 | XP_017951753.1 | 4 - 5 | 370 | 64 | ||
| GenBank mRNA | BC161565.1 | - | 4 - 5 | 334 | 64 | |
| BC170776.1 | - | 4 - 5 | 326 | 64 | ||
| BC170802.1 | - | 4 - 5 | 326 | 64 | ||
| CR760504.2 | - | 4 - 5 | 333 | 64 | ||
More Information
Set Membership: |
|

atg4a
Genomic Map