Assay Details
| Gene Symbol: fkbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016623.2 | NP_001016623.1 | 7 - 8 | 678 | 81 | |
| XM_012968431.2 | XP_012823885.1 | 7 - 8 | 726 | 81 | ||
| GenBank mRNA | BC167937.1 | - | 7 - 8 | 673 | 81 | |
| BC170855.1 | - | 7 - 8 | 669 | 81 | ||
| BC170857.1 | - | 7 - 8 | 671 | 81 | ||
| CR760490.2 | - | 7 - 8 | 678 | 81 | ||
More Information
Set Membership: |
|

fkbp3
Genomic Map