Assay Details
| Gene Symbol: rpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016626.2 | NP_001016626.1 | 1 - 2 | 107 | 81 | |
| GenBank mRNA | BC158993.1 | - | 1 - 2 | 183 | 81 | |
| BC166941.1 | - | 1 - 2 | 123 | 81 | ||
| CR760473.2 | - | 1 - 2 | 107 | 81 | ||
More Information
Set Membership: |
|

rpa3
Genomic Map