Assay Details
| Gene Symbol: nup50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016628.2 | NP_001016628.1 | 4 - 5 | 1242 | 52 | |
| GenBank mRNA | BC090603.1 | - | 4 - 5 | 1232 | 52 | |
| CR760454.2 | - | 4 - 5 | 1242 | 52 | ||
More Information
Set Membership: |
|

nup50
Genomic Map