Assay Details
| Gene Symbol: glrx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016637.2 | NP_001016637.1 | 3 - 4 | 461 | 89 | |
| XM_012961119.2 | XP_012816573.1 | 3 - 4 | 247 | 89 | ||
| XM_012961120.2 | XP_012816574.1 | 2 - 3 | 168 | 89 | ||
| GenBank mRNA | CR760424.2 | - | 3 - 4 | 461 | 89 | |
More Information
Set Membership: |
|

glrx2
Genomic Map