Assay Details
| Gene Symbol: cldn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016663.2 | NP_001016663.1 | 1 | 557 | 90 | |
| GenBank mRNA | BC167950.1 | - | 1 | 571 | 90 | |
| BC170759.1 | - | 1 | 539 | 90 | ||
| BC170761.1 | - | 1 | 539 | 90 | ||
| CR760296.2 | - | 1 | 557 | 90 | ||
More Information
Set Membership: |
|

cldn4
Genomic Map