Assay Details
| Gene Symbol: edf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016686.2 | NP_001016686.1 | 3 - 4 | 307 | 59 | |
| GenBank mRNA | CR760196.2 | - | 3 - 4 | 307 | 59 | |
More Information
Set Membership: |
|

edf1
Genomic Map