Assay Details
| Gene Symbol: stx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016709.2 | NP_001016709.1 | 7 - 8 | 732 | 71 | |
| XM_018093453.1 | XP_017948942.1 | 7 - 8 | 591 | 71 | ||
| GenBank mRNA | CR760125.2 | - | 7 - 8 | 732 | 71 | |
More Information
Set Membership: |
|

stx18
Genomic Map