Assay Details
| Gene Symbol: cat.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016716.2 | NP_001016716.1 | 714 | 92 | ||
| GenBank mRNA | CR760094.2 | - | 714 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cat.1| Gene Symbol: cat.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016716.2 | NP_001016716.1 | 714 | 92 | ||
| GenBank mRNA | CR760094.2 | - | 714 | 92 | ||
Set Membership: |
|