Assay Details
| Gene Symbol: mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016727.2 | NP_001016727.1 | 1 - 2 | 147 | 85 | |
| GenBank mRNA | BC158935.1 | - | 1 - 2 | 358 | 85 | |
| CR760046.2 | - | 1 - 2 | 147 | 85 | ||
More Information
Set Membership: |
|

mrpl43
Genomic Map