Assay Details
| Gene Symbol: cyb5r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016756.2 | NP_001016756.1 | 4 - 5 | 579 | 68 | |
| GenBank mRNA | CR855785.2 | - | 4 - 5 | 579 | 68 | |
More Information
Set Membership: |
|

cyb5r4
Genomic Map