Assay Details
| Gene Symbol: nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016790.2 | NP_001016790.1 | 11 - 12 | 1657 | 69 | |
| GenBank mRNA | CR855653.2 | - | 11 - 12 | 1657 | 69 | |
More Information
Set Membership: |
|

nup88
Genomic Map