Assay Details
| Gene Symbol: myd88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016837.2 | NP_001016837.1 | 4 - 5 | 887 | 70 | |
| GenBank mRNA | CR855486.2 | - | 4 - 5 | 887 | 70 | |
More Information
Set Membership: |
|

myd88
Genomic Map