Assay Details
| Gene Symbol: rybp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016846.2 | NP_001016846.1 | 4 - 5 | 990 | 73 | |
| GenBank mRNA | CR855445.2 | - | 4 - 5 | 990 | 73 | |
More Information
Set Membership: |
|

rybp
Genomic Map