Assay Details
| Gene Symbol: abhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016889.2 | NP_001016889.1 | 4 - 5 | 788 | 61 | |
| XM_012964753.2 | XP_012820207.1 | 4 - 5 | 1026 | 61 | ||
| XM_012964754.2 | XP_012820208.1 | 5 - 6 | 1186 | 61 | ||
| XM_012964755.2 | XP_012820209.1 | 5 - 6 | 1394 | 61 | ||
| XM_018094665.1 | XP_017950154.1 | 5 - 6 | 883 | 61 | ||
| XM_018094666.1 | XP_017950155.1 | 4 - 5 | 3811 | 61 | ||
| GenBank mRNA | BC091064.1 | - | 4 - 5 | 768 | 61 | |
| CR848568.2 | - | 4 - 5 | 788 | 61 | ||
More Information
Set Membership: |
|

abhd5
Genomic Map