Assay Details
| Gene Symbol: wls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016910.2 | NP_001016910.1 | 11 - 12 | 1556 | 73 | |
| GenBank mRNA | CR848435.2 | - | 11 - 12 | 1556 | 73 | |
More Information
Set Membership: |
|

wls
Genomic Map