Assay Details
| Gene Symbol: sphk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016918.2 | NP_001016918.1 | 2 - 3 | 635 | 69 | |
| GenBank mRNA | BC135624.1 | - | 2 - 3 | 558 | 69 | |
| CR848216.2 | - | 2 - 3 | 635 | 69 | ||
More Information
Set Membership: |
|

sphk1
Genomic Map