Assay Details
| Gene Symbol: rbmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016941.2 | NP_001016941.1 | 1 - 2 | 123 | 98 | |
| GenBank mRNA | BC159006.1 | - | 2 - 3 | 133 | 98 | |
| BC159018.1 | - | 2 - 3 | 133 | 98 | ||
| CR761911.2 | - | 1 - 2 | 123 | 98 | ||
More Information
Set Membership: |
|

rbmx2
Genomic Map