Assay Details
| Gene Symbol: tbccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016961.3 | NP_001016961.2 | 2 - 3 | 524 | 103 | |
| GenBank mRNA | BC127345.1 | - | 2 - 3 | 524 | 103 | |
| CR761361.2 | - | 2 - 3 | 626 | 103 | ||
More Information
Set Membership: |
|

tbccd1
Genomic Map