Assay Details
| Gene Symbol: stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016979.2 | NP_001016979.1 | 2 - 3 | 223 | 63 | |
| XM_012961155.2 | XP_012816609.1 | 3 - 4 | 360 | 63 | ||
| XM_012961156.2 | XP_012816610.1 | 3 - 4 | 313 | 63 | ||
| XM_012961157.2 | XP_012816611.1 | 3 - 4 | 362 | 63 | ||
| GenBank mRNA | CR848364.2 | - | 2 - 3 | 223 | 63 | |
More Information
Set Membership: |
|

stk33
Genomic Map