Assay Details
| Gene Symbol: elac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017028.2 | NP_001017028.1 | 3 - 4 | 720 | 89 | |
| GenBank mRNA | CR762044.2 | - | 3 - 4 | 720 | 89 | |
More Information
Set Membership: |
|

elac1
Genomic Map