Assay Details
| Gene Symbol: atp5pf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017042.2 | NP_001017042.1 | 2 - 3 | 210 | 89 | |
| GenBank mRNA | BC135430.1 | - | 2 - 3 | 263 | 89 | |
| CR761877.2 | - | 2 - 3 | 210 | 89 | ||
More Information
Set Membership: |
|

atp5pf
Genomic Map