Assay Details
| Gene Symbol: olfm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017043.2 | NP_001017043.1 | 3 - 4 | 414 | 101 | |
| XM_012968462.2 | XP_012823916.1 | 3 - 4 | 466 | 101 | ||
| GenBank mRNA | CR761859.2 | - | 3 - 4 | 414 | 101 | |
More Information
Set Membership: |
|

olfm4
Genomic Map