Assay Details
| Gene Symbol: ing2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017044.2 | NP_001017044.1 | 1 - 2 | 196 | 118 | |
| GenBank mRNA | BC135516.1 | - | 1 - 2 | 224 | 118 | |
| CR761858.2 | - | 1 - 2 | 196 | 118 | ||
More Information
Set Membership: |
|

ing2
Genomic Map