Assay Details
Gene Symbol: khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017045.2 | NP_001017045.1 | 1 - 2 | 98 | 63 | |
XM_012957018.2 | XP_012812472.1 | 1 - 2 | 380 | 63 | ||
GenBank mRNA | CR761855.2 | - | 1 - 2 | 98 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the CR761855.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
