Assay Details
| Gene Symbol: mrpl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017058.2 | NP_001017058.1 | 2 - 3 | 192 | 89 | |
| GenBank mRNA | BC135641.1 | - | 2 - 3 | 89 | ||
| CR761698.2 | - | 2 - 3 | 192 | 89 | ||
More Information
Set Membership: |
|

mrpl16
Genomic Map