Assay Details
| Gene Symbol: pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017072.2 | NP_001017072.1 | 1 - 2 | 100 | 74 | |
| GenBank mRNA | CR761539.2 | - | 1 - 2 | 100 | 74 | |
More Information
Set Membership: |
|

pdha1
Genomic Map