Assay Details
| Gene Symbol: leprot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017102.2 | NP_001017102.1 | 2 - 3 | 147 | 96 | |
| GenBank mRNA | CR761280.2 | - | 2 - 3 | 147 | 96 | |
More Information
Set Membership: |
|

leprot
Genomic Map