Assay Details
| Gene Symbol: zfand5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017128.2 | NP_001017128.1 | 3 - 4 | 293 | 63 | |
| XM_012963336.2 | XP_012818790.1 | 3 - 4 | 679 | 63 | ||
| XM_012963340.2 | XP_012818794.1 | 3 - 4 | 573 | 63 | ||
| GenBank mRNA | BC123970.1 | - | 3 - 4 | 287 | 63 | |
| BC166153.1 | - | 3 - 4 | 528 | 63 | ||
| CR761102.2 | - | 3 - 4 | 293 | 63 | ||
More Information
Set Membership: |
|

zfand5
Genomic Map