Assay Details
| Gene Symbol: igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017139.2 | NP_001017139.1 | 6 - 7 | 908 | 74 | |
| GenBank mRNA | BC159041.1 | - | 6 - 7 | 604 | 74 | |
| CR761008.2 | - | 6 - 7 | 908 | 74 | ||
More Information
Set Membership: |
|

igf2bp3
Genomic Map