Assay Details
| Gene Symbol: sap30l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017141.2 | NP_001017141.1 | 1 - 2 | 217 | 100 | |
| XM_012959085.2 | XP_012814539.1 | 2 - 3 | 1310 | 100 | ||
| XM_018092108.1 | XP_017947597.1 | 1 - 2 | 474 | 100 | ||
| GenBank mRNA | BC161510.1 | - | 1 - 2 | 284 | 100 | |
| CR760983.2 | - | 1 - 2 | 217 | 100 | ||
More Information
Set Membership: |
|

sap30l
Genomic Map