Assay Details
Gene Symbol: tectb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017156.2 | NP_001017156.1 | 2 - 3 | 275 | 89 | |
XM_018090235.1 | XP_017945724.1 | 2 - 3 | 524 | 89 | ||
GenBank mRNA | CR760894.2 | - | 2 - 3 | 275 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
