Assay Details
| Gene Symbol: comt.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017157.2 | NP_001017157.1 | 3 - 4 | 379 | 87 | |
| XM_012963358.2 | XP_012818812.1 | 3 - 4 | 445 | 87 | ||
| XM_012963362.2 | XP_012818816.1 | 3 - 4 | 433 | 87 | ||
| GenBank mRNA | BC153689.1 | - | 2 - 3 | 353 | 87 | |
| CR760877.2 | - | 3 - 4 | 379 | 87 | ||
More Information
Set Membership: |
|

comt.2
Genomic Map