Assay Details
| Gene Symbol: comt.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017157.2 | NP_001017157.1 | 4 - 5 | 510 | 75 | |
| XM_012963358.2 | XP_012818812.1 | 4 - 5 | 576 | 75 | ||
| XM_012963362.2 | XP_012818816.1 | 4 - 5 | 564 | 75 | ||
| GenBank mRNA | BC153689.1 | - | 3 - 4 | 484 | 75 | |
| CR760877.2 | - | 4 - 5 | 510 | 75 | ||
More Information
Set Membership: |
|

comt.2
Genomic Map