Assay Details
| Gene Symbol: gsx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017168.2 | NP_001017168.1 | 1 - 2 | 457 | 56 | |
| GenBank mRNA | BC158503.1 | - | 1 - 2 | 440 | 56 | |
| CR855836.2 | - | 1 - 2 | 457 | 56 | ||
| DQ195531.1 | - | 1 - 2 | 478 | 56 | ||
More Information
Set Membership: |
|

gsx2
Genomic Map