Assay Details
| Gene Symbol: cenpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017175.2 | NP_001017175.1 | 5 - 6 | 457 | 74 | |
| GenBank mRNA | BC135574.1 | - | 5 - 6 | 475 | 74 | |
| CR760711.2 | - | 5 - 6 | 457 | 74 | ||
More Information
Set Membership: |
|

cenpm
Genomic Map