Assay Details
| Gene Symbol: rdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017189.2 | NP_001017189.1 | 4 - 5 | 782 | 72 | |
| XM_012957042.2 | XP_012812496.1 | 4 - 5 | 962 | 72 | ||
| GenBank mRNA | CR760619.2 | - | 4 - 5 | 782 | 72 | |
More Information
Set Membership: |
|

rdh7
Genomic Map