Assay Details
| Gene Symbol: scamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017190.2 | NP_001017190.1 | 7 - 8 | 766 | 60 | |
| GenBank mRNA | BC167666.1 | - | 7 - 8 | 787 | 60 | |
| CR760611.2 | - | 7 - 8 | 766 | 60 | ||
More Information
Set Membership: |
|

scamp1
Genomic Map