Assay Details
| Gene Symbol: tomm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017214.2 | NP_001017214.1 | 105 | 77 | ||
| GenBank mRNA | CR760426.2 | - | 105 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tomm7| Gene Symbol: tomm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017214.2 | NP_001017214.1 | 105 | 77 | ||
| GenBank mRNA | CR760426.2 | - | 105 | 77 | ||
Set Membership: |
|