Assay Details
| Gene Symbol: pik3r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017229.2 | NP_001017229.1 | 2 - 3 | 449 | 79 | |
| GenBank mRNA | BC136130.1 | - | 2 - 3 | 463 | 79 | |
| BC166070.1 | - | 2 - 3 | 452 | 79 | ||
| CR760256.2 | - | 2 - 3 | 449 | 79 | ||
More Information
Set Membership: |
|

pik3r3
Genomic Map